Specifications
- Antibody Type:Primary
- Antigen Name:mucin 1, cell surface associated
- Antigen Symbol:MUC1
- Clonality:Monoclonal
- Clone:MUC1/967
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P15941
- Antigen Synonyms:MUC1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human milk fat globule membranes were used as the immunogen for this Epithelial Marker Antigen antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-736
Specifications
About this item
This Epithelial Membrane Antigen / EMA antibody, also called MUC1 and Mucin-1, recognizes the full-length protein in a glycosylation-independent manner and can bind to the fully glycosylated protein. The dominant epitope of this mAb is APDTR in the VNTR region. It reacts with the core peptide of the EMA protein, which is a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (>200kDa) and an amino acid composition rich in serine, threonine, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats that functions as multiple epitopes for the mAb. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in staining intensity between normal and malignant tissues. This EMA antibody reacts with both normal and malignant epithelia of various tissues including breast and colon.
Flow Cytometry: 0.5-1 ug/million cells IF: 1-2 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Epithelial Marker Antigen antibody to be titered up or down for optimal performance.
1. FFPE staining REQUIRES boiling sections in 10mM citrate buffer, pH 6, for 10-20 min followed by cooling at RT for 10-20 min.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/967
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human