Specifications
- Antibody Type:Primary
- Antigen Name:integrin, beta 4
- Antigen Symbol:ITGB4
- Clonality:Monoclonal
- Clone:UM-A9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P16144
- Antigen Synonyms:ITGB4
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:The human squamous cell carcinoma line (UM-SCC1) was used as the immunogen for this Integrin beta-4 / CD104 antibody.
- Tested Applications:Blocking
- Purification:Protein G affinity chromatography
- Cat. No.:76195-540
Specifications
About this item
This antibody recognizes a protein of 205kDa, which is identified as integrin beta 4 (ITGB4) and CD104. Its epitope is localized in the extracellular domain of integrin beta 4 protein. Integrins are heterodimers comprised of a and b subunits, that are non-covalently associated transmembrane glycoprotein receptors. Different combinations of a and b polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha-6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Functional Blocking: partially blocks laminin binding; order the BSA/azide free format Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: UM-A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human