Order Entry
United States
ContactUsLinkComponent
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK1]
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK1]
Catalog # 76193-872
Supplier:  Prosci
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK1]
Catalog # 76193-872
Supplier:  Prosci
Supplier Number:  33-141

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    heat shock 60kDa protein 1 (chaperonin)
  • Antigen Symbol:
    HSPD1
  • Clonality:
    Monoclonal
  • Clone:
    LK1
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rat,
    Mouse,
    Hamster,
    Drosophila,
    Pig,
    Rabbit,
    Monkey,
    Xenopus,
    Sheep,
    Chicken,
    Human,
    Bovine,
    Dog
  • Western Blot:
    Yes
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P10809
  • Antigen Synonyms:
    HSPD1
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant human HSP60 was used as the immunogen for this antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76193-872

Specifications

About this item

This antibody recognizes a 60kDa protein, identified as the heat shock protein 60. The antibody's epitope is localized between amino acids 383-447 of human HSP60. A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp’s), more appropriately called as stress response proteins (srp’s). HSP60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial HSP60, but also against its mammalian homolog. Clone LK1 antibody, unlike LK2 antibody, recognizes only the mammalian (not bacterial) HSP60 and is useful in distinguishing HSP60 from mammals and bacteria.

Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml WB: 0.25-0.5 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat, Hamster, Sheep, Rabbit, Bovine, Dog, Pig, Monkey, Chicken, Xenopus, Drosophila