Order Entry
United States
ContactUsLinkComponent
Anti-CD1A Mouse Monoclonal Antibody [clone: C1A/711]
Anti-CD1A Mouse Monoclonal Antibody [clone: C1A/711]
Catalog # 76193-816
Supplier:  Prosci
Anti-CD1A Mouse Monoclonal Antibody [clone: C1A/711]
Catalog # 76193-816
Supplier:  Prosci
Supplier Number:  33-077

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD1a molecule
  • Antigen Symbol:
    CD1A
  • Clonality:
    Monoclonal
  • Clone:
    C1A/711
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P06126
  • Antigen Synonyms:
    CD1A
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant human CD1a protein was used as the immunogen for this antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76193-816

Specifications

About this item

At least five CD1 genes (CD1a, b, c, d, and e) are identified. CD1 proteins have been demonstrated to restrict T cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation. CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta-2 microglobulin. CD1a antibody labels Langerhans cell histiocytosis (Histiocytosis X), extranodal histiocytic sarcoma, a subset of T-lymphoblastic lymphoma/leukemia, and interdigitating dendritic cell sarcoma of the lymph node. When combined with antibody against TTF-1 and CD5, CD1a antibody is useful in distinguishing between pulmonary and thymic neoplasms since CD1a is consistently expressed in thymic lymphocytes in both typical and atypical thymomas, but only focally in 1/6 of thymic carcinomas and not in lymphocytes in pulmonary neoplasms. antibody to CD1a is reported to be a new marker for perivascular epithelial cell tumor (PEComa).

Flow Cytometry: 0.5-1 ug/million cells IF: 1-2 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: C1A/711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human