Specifications
- Antibody Type:Primary
- Antigen Name:cyclin D1
- Antigen Symbol:CCND1
- Clonality:Monoclonal
- Clone:DCS-6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P24385
- Antigen Synonyms:CCND1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human recombinant full length protein was used as the immunogen for this Cyclin D1 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-422
Specifications
About this item
This antibody recognizes a protein of 36kDa, identified as Cyclin D1. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms and a key cell cycle regulator. This antibody neutralizes the activity of Cyclin D1 in vivo. About 60% of mantle cell lymphomas (MCL) contain a t(11; 14)(q13; q32) translocation resulting in over-expression. Cyclin D1 antibody is useful in identifying mantle cell lymphomas, which stain positive, from CLL/SLL and follicular lymphomas, which stain negative. Occasionally, hairy cell leukemia and plasma cell myeloma weakly express Cyclin D1.
- Flow Cytometry: 0.5-1 ug/million cells IF: 1-2 ug/ml WB: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Cyclin D1 antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
- 2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: DCS-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Monkey, Mouse, Rat