About this item
Teach a variety of skills with just one injection simulator.
- Safe for use with air or fluid injection training
- Useable with multiple needle sizes and lengths
- Designed to withstand thousands of punctures
PRACTI-Injecta Pad® injection simulator is designed to withstand thousands of needle punctures as your students practice injection techniques. Available in 2 sizes (Large 7×7×2.5 and Small 5×5×2.5) the 2.5 inch thickness allows for needles of multiple lengths to be used. Wallcur’s PRACTI-Injecta Pad® is the only injection simulator that perfectly simulates aspiration tension, so vital when teaching proper I.M. injection techniques.
Intramuscular, subcutaneous, and intradermal injections
Z-track and many other injection techniques
Tissue displacement skills
Skin puncture resistance
Aspiration tension
Also available is a Sim-Skin pad. This 3×3 pad is for training for subcutaneous injections. This skin reacts like real skin forming a 'bleb' or 'wheal' upon successful injection.