Order Entry
United States
ContactUsLinkComponent
D-(-)-Luciferin potassium salt ≥99.5% endotoxin-free
D-(-)-Luciferin potassium salt ≥99.5% endotoxin-free
Catalog # 103258-192
Supplier:  OZ Biosciences
CAS Number:  
D-(-)-Luciferin potassium salt ≥99.5% endotoxin-free
Catalog # 103258-192
Supplier:  OZ Biosciences
Supplier Number:  LK10000
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    103258-192
  • Supplier no.:
    LK10000

Specifications

About this item

 :
 :318.41 g/mol
CAS Number:115144-35-9
Formula:C₁₁H₇KN₂O₃S₂
MDL Number:MFCD00044928