Order Entry
United States
ContactUsLinkComponent
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
Catalog # 103258-194
Supplier:  OZ Biosciences
CAS Number:  
undefined
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
Catalog # 103258-194
Supplier:  OZ Biosciences
Supplier Number:  LN10000
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    103258-194
  • Supplier no.:
    LN10000

Specifications

About this item

 :
 :302.30 g/mol
CAS Number:103404-75-7
Formula:C₁₁H₇N₂NaO₃S₂
MDL Number:MFCD00044938