Order Entry
United States
ContactUsLinkComponent
G418 disulfate, Molecular Biology Grade
G418 disulfate, Molecular Biology Grade
Catalog # 103258-196
Supplier:  OZ Biosciences
CAS Number:  
G418 disulfate, Molecular Biology Grade
Catalog # 103258-196
Supplier:  OZ Biosciences
Supplier Number:  GS21000
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    103258-196
  • Supplier no.:
    GS21000

Specifications

About this item

 :
 :692.72 g/mol
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314