


Aquaphenol™, clear, colorless solution
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
Density:1.03 g/cm³ (20 °C) (lit.)
UN:2821
ADR:6.1,III
:GHS05
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Iron:≤0.0001%
- Heavy Metals:≤0.0005%
- Water Content (KF):~30%
- RNase, DNase, Protease:Not Detected
Specification Test Results
Frequently Bought Together