Order Entry
United States
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
Irritant
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
Catalog # 76020-286
Supplier:  MP Biomedicals
CAS Number:  
undefined
Irritant
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
Catalog # 76020-286
Supplier:  MP Biomedicals
Supplier Number:  114064112
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Size:
    5 g
  • Cat. No.:
    76020-286

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • pH (4% aq):
    5.0-7.0
  • Optical Rotation (c=1, water):
    -26° to -33°

Specification Test Results

About this item

 :
 :238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:105…115 °C
EINECS:206-703-0
 :GHS07
Storage Temperature:Freezer
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273