ContactUsLinkComponent
Myoblast Differentiation Medium, iXCells Biotechnologies
Myoblast Differentiation Medium, iXCells Biotechnologies
Catalog # 103521-492
CAS Number:  
Myoblast Differentiation Medium, iXCells Biotechnologies
Catalog # 103521-492
Supplier Number:  MD-0065-500ML
CAS Number:  
Specifications
  • Media Format:
    Liquid
  • Size:
    500 ml
  • Cat. No.:
    103521-492
Specifications
About this item
Myoblast Differentiation Medium; 500 mL; Myoblast Differentiation Medium is designed to promote myoblasts differentiation in vitro
This medium is fully supplemented with fetal bovine serum, growth factors without antibiotic.