Mouse Keratinocyte Differentiation Medium, iXCells Biotechnologies
Catalog # 103521-488
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Media Format:Liquid
- Size:500 ml
- Cat. No.:103521-488
Specifications
About this item
Mouse Keratinocyte Differentiation Medium is designed to promote mouse keratinocytes differentiation in vitro
This medium is fully supplemented with fetal bovine serum, growth factors without antibiotic.