Rat Astrocytes (RA), iXCells Biotechnologies
Catalog # 103521-250
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-250
Specifications
About this item
Rat Astrocytes (RA); Frozen
- High-quality Primary Rat Astrocytes (RA)
- High purity
RA are isolated from postnatal day 2 rat brain. RA are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast and fungi.