Mouse Brain Vascular Pericytes (MBVP), iXCells Biotechnologies
Catalog # 103521-248
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-248
Specifications
About this item
Mouse Brain Vascular Pericytes (MBVP); Frozen
- High-quality Primary Mouse Brain Vascular Pericytes (MBVP)
- High purity
MBVP are isolated from adult mouse brain. MBVP are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast and fungi.