ContactUsLinkComponent
Human Peripheral Blood CD4+ Cells (Helper T Cells), iXCells Biotechnologies
Human Peripheral Blood CD4+ Cells (Helper T Cells), iXCells Biotechnologies
Catalog # 103521-816
CAS Number:  
Human Peripheral Blood CD4+ Cells (Helper T Cells), iXCells Biotechnologies
Catalog # 103521-816
Supplier Number:  10HU-023-10M
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Size:
    10 million cells/vial
  • Cat. No.:
    103521-816
Specifications
About this item
Human Peripheral Blood CD4+ Cells (Helper T Cells); Frozen
  • High-quality Human Peripheral Blood CD4+ Cells (Helper T Cells)
  • High purity
  • IRB-approved
Human Peripheral Blood CD4+ Helper T Cells are isolated from human peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic selection. > 93% of the cells are CD4 positive.