Human Hepatic Sinusoidal Endothelial Cells (HHSEC), iXCells Biotechnologies
103521-204
:
- Cell Format:Cryopreserved
- Size:0.5 million cells/vial
- Cat. No.:103521-204
Human Hepatic Sinusoidal Endothelial Cells (HHSEC); Frozen; 0.5 million cells/vial
- High-quality primary Human Hepatic Sinusoidal Endothelial Cells
- High purity
- IRB-approved
HHSEC are isolated from human liver. HHSEC are characterized by CD31 and vWF/Factor VIII staining. HHSEC are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.