ContactUsLinkComponent
Human Primary Astrocytes, iXCells Biotechnologies
Catalog # 103521-792
CAS Number:  
undefined
Human Primary Astrocytes, iXCells Biotechnologies
Catalog # 103521-792
Supplier Number:  10HU-035
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Size:
    0.5 million cells/vial
  • Cat. No.:
    103521-792
Specifications
About this item
Human Astrocytes (HA); Frozen
  • High-quality primary Human Astrocytes
  • High purity
  • IRB-approved
HA are isolated from human brain (cerebral cortex). HA are cryopreserved at passage one and delivered frozen. HA are characterized by immunofluorescence with antibody specific to GFAP. Negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.