Order Entry
United States
ContactUsLinkComponent
SensoLyte® 520 FRET SIRT1 Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 FRET SIRT1 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-514
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 FRET SIRT1 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-514
Supplier:  Anaspec
Supplier Number:  AS-72155
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-514
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72155

Specifications

About this item

Histone deacetylase (HDAC) enzymes act as transcriptional repressors of genes through the deacetylation of lysine residues on histone proteins

The SensoLyte® 520 FRET SIRT1 Assay Kit provides a convenient, two-step homogeneous procedure for measuring sirtuin 1 activity using FRET substrate. In the first step, an acetylated substrate is incubated with SIRT1 containing samples. Deacetylation of substrate first sensitizes it, so that, in the second step mixing with the SIRT developer generates a fluorophore, which can then be detected at Ex/Em= 490/520 nm. This kit can be used for high throughput screening of SIRT1 inhibitors/activators. The long wavelength fluorescence of FRET substrate is less interfered by the autofluorescence of cell components and test compounds. The nuclear extract HDAC (containing HDACs 1 and 2) does not deacetylate FRET sirtuin substrate. Assays are performed in a standard 96-well format.

Sirtuins (SIRTs) are a unique class of deacetylases (class III HDACs) that require NAD as a cofactor. Sirtuin 1 (SIRT1) is involved in regulation of tumor suppressor p53 and inhibition of apoptosis. It also represents a target for treatment of age-related diseases and type II diabetes.