Order Entry
United States
ContactUsLinkComponent
SensoLyte® AFC Urokinase (uPA) Activity Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® AFC Urokinase (uPA) Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-522
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® AFC Urokinase (uPA) Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-522
Supplier:  Anaspec Inc
Supplier Number:  AS-72159
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-522
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72159

Specifications

About this item

Urokinase-type plasminogen activator (uPA) is a serine protease that functions in the conversion of the zymogen plasminogen to the active, broad-spectrum serine protease plasmin

The SensoLyte® AFC Urokinase (uPA) Assay Kit is optimized for screening of enzyme inhibitors. This kit contains a fluorogenic substrate with a high reactivity and low background. uPA cleaves the substrate resulting in release of AFC (7-amino-4-trifluoromethylcoumarin) fluorophore. Fluorescence can be monitored at excitation /emission= 380/500 nm.

Plasmin, in turn, mediates the pericellular proteolysis of extracellular matrix components and activates other proteases such as matrix metalloproteinases and collagenases that lead to the further degradation and remodeling of the extracellular matrix. Besides the proteolytic function, upon binding to uPA receptor (uPAR), uPA is involved in initiating versatile intracellular signal pathways that regulate cell proliferation, adhesion, and migration through its interaction with various integrins and vitronectin. uPA has been implicated in a number of normal and pathological processes, and recognized as a prognostic marker for various cancers as well as a target for drug discovery.