Order Entry
United States
ContactUsLinkComponent
SensoLyte® 520 Factor Xa Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 Factor Xa Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-624
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 Factor Xa Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-624
Supplier:  Anaspec
Supplier Number:  AS-72208
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-624
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72208

Specifications

About this item

Factor Xa (FXa) is a serine endopeptidase composed of two disulfide-linked subunits

The SensoLyte® 520 Factor Xa Assay Kit provides a convenient assay for high throughput screening of FXa inhibitors or continuous assay of enzyme activity. It detects FXa activity using a 5-FAM/QXL® 520 fluorescence resonance energy transfer (FRET) peptide. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage into two separate fragments by FXa, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission wavelengths = 490 nm/520 nm. With excellent fluorescence quantum yield and longer wavelength, 5-FAM shows less interference from autofluorescence of test compounds and cellular components. The assays are performed in a convenient 96-well microplate format.

FXa leads to blood clot formation by converting prothrombin to thrombin through the prothrombinase complex. FXa is generated from zymogen Factor X via the intrinsic and extrinsic pathways and is the rate-limiting step in the propagation of thrombin generation. In the presence of Ca2+ ions, FXa forms prothrombinase with factor Va on the phospholipid membrane of the activated platelets. FXa has emerged as an attractive target for drug discovery for thromboembolic diseases.