Specifications
- Size:Two 96-well Plate
- Cat. No.:103010-164
- Storage temperature:–20 °C
- Includes:Two 96-well plates, essential components and detailed protocol and references
- Supplier no.:AS-71136
Specifications
About this item
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
SensoLyte® 520 MMP Profiling Kit contains a series of well-designed FRET peptide substrates for use as fluorogenic indicators to assay MMP protease activities. It provides the best solution for profiling MMPs or MMP inhibitors.
They are involved in diseases such as cancer, arthritis, asthma, angiogenesis, alopecia and metastasis. They are also identified as drug-screening targets.