Avantor® Alltima, AQ HPLC Columns
Supplier: Avantor
Certificates
About this item
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
Specifications
- Catalog No:
- 75797-164
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001:2015 (530840 UM15)