- Description:24 Place SEM Holder
- Cat. No.:103306-402
- Supplier no.:75958-67
This 24 place SEM holder perfect with Hitachi VP SEM.
- Made of aluminum
- Unique ID numbers for each hole
This 24-place holder offers a stage travel of 100 x 50 mm. The holder measures 4 7/16 L x 21/2 W x 3/8" H (112.7 x 63.5 x 10 mm) and has a 1/8" (3.2 mm) mounting pin.