These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
Style 7B tweezers with curved tips and inside serrations. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox. Resistant up to temperatures of 400 °C (DIN 50 914) and suitable for autoclave sterilization at 270 °C.