ContactUsLinkComponent
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
  103302-376
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
  103302-376
 :  75954-01
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Mini SEM Clamp
  • Environmentally Preferable:
  • Cat. No.:
    103302-376
  • Supplier no.:
    75954-01
  • Opening size:
    12.7 mm
Clamp holder used for flat or thin specimens for Scanning Electron Microscopy.
  • Uses standard 12.7 mm SEM pin stubs
  • Vacuum grade aluminum with brass screws
  • Use for silicon chips, foil, wires, and small tubes
Suitable for holding flat or thin specimens such as silicon chips, foil, wires, and small tubes. Made of Vacuum-grade aluminum with brass screw.