Rat IgG2b Isotype Control [clone: LTF-2]
75842-448
: Biogems
- Antibody Type:Control
- Antigen Name:IgG2b Isotype Control (LTF-2)
- Clonality:Monoclonal
- Clone:LTF-2
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Cat. No.:75842-448
- Supplier no.:41823-25-500
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse