Rat IgG2a Isotype Control [clone: 2A3]
75844-508
: Biogems
- Antibody Type:Control
- Antigen Name:IgG2a Isotype Control (2A3)
- Clonality:Monoclonal
- Clone:2A3
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Rat
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Cat. No.:75844-508
- Supplier no.:41813-25-100
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rat