Rat IgG1 Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HRPN]
75842-476
: Biogems
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-476
- Supplier no.:42813-70-100
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat