Rat IgG2b Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: LTF-2]
Catalog # 75842-458
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75842-458
- Supplier no.:41823-70-100
Specifications
About this item
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse