Order Entry
United States
ContactUsLinkComponent
Rat IgG2a Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 2A3]
Rat IgG2a Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 2A3]
Catalog # 75842-436
Supplier:  Biogems
Rat IgG2a Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 2A3]
Catalog # 75842-436
Supplier:  Biogems
Supplier Number:  41813-70-100

Specifications

  • Antibody Type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    2A3
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Form:
    liquid
  • Cat. No.:
    75842-436
  • Supplier no.:
    41813-70-100

Specifications

About this item

The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rat