Mouse IgG1, kappa Isotype Control (PE (Phycoerythrin)) [clone: MOPC-21]
75841-348
: Biogems
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:100ug
- Form:liquid
- Cat. No.:75841-348
- Supplier no.:44212-60-100
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse