Rat IgG2b, kappa Isotype Control (PE (Phycoerythrin)) [clone: LTF-2]
Catalog # 75842-454
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-454
- Supplier no.:41823-60-100
Specifications
About this item
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Rat