Order Entry
United States
ContactUsLinkComponent
 
Armenian Hamster IgG Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: 299Arm]
Catalog # 75841-356
Supplier:  Biogems
Armenian Hamster IgG Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: 299Arm]
Catalog # 75841-356
Supplier:  Biogems
Supplier Number:  44213-50-50

Specifications

  • Antibody Type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    299Arm
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Armenian hamster
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Size:
    50ug
  • Form:
    liquid
  • Cat. No.:
    75841-356
  • Supplier no.:
    44213-50-50

Specifications

About this item

The 299Arm monoclonal antibody is used as an Isotype Control immunoglobulin for Armenian hamster IgG antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: 299Arm
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: mouse