Mouse IgG1 Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: MOPC-21]
Catalog # 75841-346
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75841-346
- Supplier no.:44212-50-100
Specifications
About this item
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse