Mouse IgG2b Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: MPC-11]
Catalog # 75841-330
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75841-330
- Supplier no.:43922-50-100
Specifications
About this item
The MPC-11 monoclonal antibody is expressed by a cell line adapted from the Merwin Plasmacytoma-11, and has unknown specificity. The MPC-11 line was obtained by intraperitoneal implantation into a BALB/c Mouseof a Millipore diffusion chamber and is used as an Isotype Control for MouseIgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse