Rat IgG1 Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: HRPN]
Catalog # 75842-472
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75842-472
- Supplier no.:42813-50-100
Specifications
About this item
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse