Rat IgG2b Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: LTF-2]
75842-452
: Biogems
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-452
- Supplier no.:41823-50-100
The RatG2b monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat