Rat IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: 2A3]
Catalog # 75842-430
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-430
- Supplier no.:41813-50-100
Specifications
About this item
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rat