Order Entry
United States
ContactUsLinkComponent
Mouse IgG1 Isotype Control (BG Violet 450) [clone: MOPC-21]
Mouse IgG1 Isotype Control (BG Violet 450) [clone: MOPC-21]
Catalog # 75841-344
Supplier:  Biogems
Mouse IgG1 Isotype Control (BG Violet 450) [clone: MOPC-21]
Catalog # 75841-344
Supplier:  Biogems
Supplier Number:  44212-40-100

Specifications

  • Antibody Type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    BG Violet 450
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Size:
    100 μg
  • Form:
    liquid
  • Cat. No.:
    75841-344
  • Supplier no.:
    44212-40-100

Specifications

About this item

MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse