Rat IgG1 Isotype Control (APC (Allophycocyanin)) [clone: HRPN]
75842-480
: Biogems
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-480
- Supplier no.:42813-80-100
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat