Rat IgG2b Isotype Control (APC (Allophycocyanin)) [clone: LTF-2]
Catalog # 75842-462
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-462
- Supplier no.:41823-80-100
Specifications
About this item
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat