Rat IgG1 Isotype Control (PE (Phycoerythrin)-Cy7®) [clone: HRPN]
Catalog # 75842-478
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Rat
- Size:100 μg
- Form:liquid
- Cat. No.:75842-478
- Supplier no.:42813-77-100
Specifications
About this item
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat