Order Entry
United States
ContactUsLinkComponent
Rat IgG2b Isotype Control (PE (Phycoerythrin)-Cy5®) [clone: LTF-2]
Rat IgG2b Isotype Control (PE (Phycoerythrin)-Cy5®) [clone: LTF-2]
  75842-456
 :  Biogems
Rat IgG2b Isotype Control (PE (Phycoerythrin)-Cy5®) [clone: LTF-2]
  75842-456
 :  Biogems
 :  41823-65-100

 

  • Antibody Type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    LTF-2
  • Conjugation:
    PE (Phycoerythrin)-Cy5®
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    100 μg
  • Form:
    liquid
  • Cat. No.:
    75842-456
  • Supplier no.:
    41823-65-100

 

 

The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse