Mouse IgG2a Isotype Control (PE (Phycoerythrin)-Cy7®) [clone: C1.18.4]
Catalog # 75843-380
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75843-380
- Supplier no.:43912-77-100
Specifications
About this item
The C1.18.4 monoclonal antibody is used as an Isotype Control for MouseIgG2a antibodies. It has unknown specificity and occurred spontaneously in a C3H mouse.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse