Mouse IgG1 Isotype Control (PE (Phycoerythrin)-Cy7®) [clone: MOPC-21]
Catalog # 75841-350
Supplier: Biogems
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75841-350
- Supplier no.:44212-77-100
Specifications
About this item
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse