Anti-TLR2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TL2.1]
Catalog # 75843-232
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Name:Toll-like receptor 2
- Antigen Symbol:TLR2
- Clonality:Monoclonal
- Clone:TL2.1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:TLR-2|toll-like receptor 2
- Cat. No.:75843-232
- Supplier no.:24911-50-100
Specifications
About this item
The TL2.1 monoclonal antibody specifically reacts with human CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on peripheral blood monocytes and is involved distinguishing bacterial lipoproteins. The TL2.1 antibody blocks the receptor and immunoprecipitates TLR2 from human mammary epithelial cells (HMEC) and peripheral blood mononuclear cells (PBMC).
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: TL2.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human