Anti-MHCI Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 34-1-2S]
75841-426
: Biogems
- Antibody Type:Primary
- Antigen Name:MHC Class I
- Antigen Symbol:MHCI
- Clonality:Monoclonal
- Clone:34-1-2S
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:50 μg
- Form:liquid
- Antigen Synonyms:H2Kd|MHCI|H2Dd
- Cat. No.:75841-426
- Supplier no.:86112-50-50
The 34-1-2S monoclonal antibody specifically reacts with the mouse H-2k/H2D MHC class I alloantigens, which are involved in antigen presentation to T cells. The 34-1-2S antibody cross-reacts with the b, p, q, r, and s haplotypes.
Type: Primary
Antigen: MHCI
Clonality: Monoclonal
Clone: 34-1-2S
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse