Order Entry
United States
ContactUsLinkComponent
 
Anti-IL-4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP4-25D2]
  75843-532
 :  Biogems
Anti-IL-4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP4-25D2]
  75843-532
 :  Biogems
 :  81121-80-25

 

  • Antibody Type:
    Primary
  • Antigen Name:
    IL-4
  • Antigen Symbol:
    IL-4
  • Clonality:
    Monoclonal
  • Clone:
    MP4-25D2
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    25 tests
  • Form:
    liquid
  • Antigen Synonyms:
    IL4|B-cell stimulating factor-1|Interleukin-4
  • Cat. No.:
    75843-532
  • Supplier no.:
    81121-80-25

 

 

The MP4-25D2 monoclonal antibody specifically reacts with human Interleukin-4 (IL-4), expressed by the activated T cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen. The MP4-25D2 is a neutralizing antibody and is reported to be cross-reactive with rhesus monkeys.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Human