- Antibody Type:Primary
- Antigen Symbol:CD90.2
- Clonality:Monoclonal
- Clone:30-H12
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:25 μg
- Form:liquid
- Antigen Synonyms:Thy-1.2
- Cat. No.:75842-786
- Supplier no.:03012-50-25
The 30-H12 monoclonal antibody specifically binds to mouse CD90.2, an alloantigen known as Thy-1.2, expressed on thymocytes, mature T cells, epithelial cells, neurons, hematopoietic stem cells, and fibroblasts. CD90 is a membrane glycoprotein that regulates the adhesion and signal transduction in T lymphocytes, and the adhesion of thymocytes to thymic stroma. The interaction between 30-H12 and the antibody to the CD3/TCR complex upregulates thymocytes signal transduction and apoptosis and downregulates mature T cell proliferation. The 30-H12 antibody seems to be unable to cross-link with CD90.1.
Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse